Human Recombinant TGF-beta 1 (from HEK293 Cells), Biotin
103871-632
:
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Tag sequence:Avitag™
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Biotinylated Human TGF-Beta 1, Avitag™;;;;;;;
- Gene ID:P01137-1
- Protein synonyms:TGF-beta-1|CEDLAP|TGF-beta 1|TGFB|DPD1|TGFB1
- Protein/peptide name:TGF-beta 1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:14.5 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 60% ACN, 0.085% TFA in Water. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-632
Biotinylated Human TGF-Beta 1 / TGFB1 Protein, Avitag™, ACROBiosystems