Human Recombinant NKG2D (from HEK293 Cells)
Catalog # 103871-576
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 0.1 EU per μg by the LAL method.
- Gene ID:P26718-1
- Protein synonyms:KLRK1|NK cell receptor D|CD314|NKG2D
- Protein/peptide name:NKG2D
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:43.5 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-576
Specifications
About this item
Human NKG2D / CD314 Protein, Mouse IgG2a Fc Tag, ACROBiosystems