Human Recombinant Mucin-1 (from HEK293 Cells)
Catalog # 103815-168
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P15941-1
- Protein synonyms:Mucin 1|Episialin|CA 15-3|PEMT|PUM|MUC-1/X|CD227|MUC-1/SEC|KL-6|EMA|MUC1|MAM6|PEM|H23AG|MUC1/ZD
- Protein/peptide name:Mucin-1
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:21.3 kDa (partial α chain) & 8.4 kDa (partial β chain)
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103815-168
Specifications
About this item
Human Mucin-1 / MUC-1 (890-1158) Protein, His Tag, ACROBiosystems