Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Mucin-1 (from HEK293 Cells)
Human Recombinant Mucin-1 (from HEK293 Cells)
Catalog # 103815-152
CAS Number:  
Human Recombinant Mucin-1 (from HEK293 Cells)
Catalog # 103815-152
Supplier Number:  MU1H52H5100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P15941-1
  • Protein synonyms:
    Mucin 1|Episialin|CA 15-3|PEMT|PUM|MUC-1/X|CD227|MUC-1/SEC|KL-6|EMA|MUC1|MAM6|PEM|H23AG|MUC1/ZD
  • Protein/peptide name:
    Mucin-1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    7.2 kDa (partial α chain) & 8.2 kDa (partial β chain)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103815-152

Specifications

About this item

Human Mucin-1 / MUC-1 (1036-1155) Protein, His Tag, ACROBiosystems