Human Recombinant MAG/ Siglec-4a (from HEK293 Cells)
Catalog # 103815-230
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P20916-1
- Protein synonyms:GMA|MAG|Siglec-4a|S-MAG
- Protein/peptide name:MAG/ Siglec-4a
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:81.1 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103815-230
Specifications
About this item
Human MAG / Siglec-4a Protein, Fc Tag, ACROBiosystems