Human Recombinant LILRB4 (from HEK293 Cells)
Catalog # 103815-328
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Tag sequence:His Tag (SPR verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) immobilized on CM5 Chip can bind Human LILRB4, His Tag (SPR verified) (Cat. No. LI4-H52H7) with an affinity constant of 4.67 μM as determined in a SPR assay (Biacore T200) (QC tested).
- Protein synonyms:CD85K|HM18|ILT3|LIR5|LILRB4
- Protein/peptide name:LILRB4
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:28.2 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103815-328
Specifications
About this item
Human LILRB4 / CD85k / ILT3 Protein, His Tag (SPR verified), ACROBiosystems