Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-8 (from HEK293 Cells)
Human Recombinant IL-8 (from HEK293 Cells)
Catalog # 103871-524
CAS Number:  
Human Recombinant IL-8 (from HEK293 Cells)
Catalog # 103871-524
Supplier Number:  IL8H5258100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P10145-1
  • Protein synonyms:
    MDNCF|LYNAP|IL8|NAP-1|GCP1|MONAP|LECT|NAF|CXCL8|LUCT
  • Protein/peptide name:
    IL-8
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    34.8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-524

Specifications

About this item

Human IL-8 / CXCL8 Protein, Fc Tag, ACROBiosystems