Human Recombinant IL-2 R beta (from HEK293 cells)
Catalog # 103814-984
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Tag sequence:His, Avitag™
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Loaded Biotinylated Human IL-2 R beta, His,Avitag (Cat. No. ILB-H82E3) on SA Biosensor, can bind Human IL-2, Tag Free (Cat. No. IL2-H4113) with an affinity constant of 0.71 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested). Loaded Biotinylated Human IL-2 R beta, His,Avitag (Cat. No. ILB-H82E3) on SA Biosensor, can bind Human IL-15, Tag Free (Cat. No. IL5-H4117) with an affinity constant of 16.5 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P14784-1
- Protein synonyms:CD122|P70-75|IL2RB|RP5-1170K4.6
- Protein/peptide name:IL-2 R beta
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:28.2 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103814-984
Specifications
About this item
Biotinylated Human IL-2 R beta / CD122 Protein, His,Avitag™, ACROBiosystems