Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-13 R alpha 2 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Human Recombinant IL-13 R alpha 2 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
  103814-892
 :  
Human Recombinant IL-13 R alpha 2 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
  103814-892
 :  IL2HF2H3200UG
 :  

 

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    200 µg
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 ℃, aviod light
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human IL-13 R alpha 2, His Tag (Cat. No. IL2-HF2H3) with a linear range of 0.313-1.25 μg/mL (QC tested).
  • Gene ID:
    Q14627-1
  • Protein synonyms:
    IL13RA2|CD213A2|IL-13R|CT19|IL13BP
  • Protein/peptide name:
    IL-13 R alpha 2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    39.0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103814-892

 

 

FITC-Labeled Human IL-13 R alpha 2 Protein, His Tag, ACROBiosystems