Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant IgG1 Fc (from HEK293 cells)
  103815-036
 :  
Human Recombinant IgG1 Fc (from HEK293 cells)
  103815-036
 :  IG1H52C9500UG
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    500 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P01857-1 (C103S)
  • Protein synonyms:
    IgG1
  • Protein/peptide name:
    IgG1 Fc
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    27.1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103815-036

 

 

Human IgG1 Fc Protein, Flag Tag, ACROBiosystems