Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Her2 (from HEK293 cells)
Human Recombinant Her2 (from HEK293 cells)
Catalog # 103871-620
CAS Number:  
Human Recombinant Her2 (from HEK293 cells)
Catalog # 103871-620
Supplier Number:  HE2H52H4100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P04626-1
  • Protein synonyms:
    NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
  • Protein/peptide name:
    Her2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    18.5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-620

Specifications

About this item

Human Her2 / ErbB2 (498-648) Protein, His Tag, ACROBiosystems