Order Entry
Canada
ContactUsLinkComponent
Human Recombinant GUCY2C (from HEK293 cells)
Human Recombinant GUCY2C (from HEK293 cells)
Catalog # 103871-718
CAS Number:  
Human Recombinant GUCY2C (from HEK293 cells)
Catalog # 103871-718
Supplier Number:  GUCH52H5100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    2e5 of anti-GUCY2C CAR-293 cells were stained with 100 µL of 1 µg/mL Human GUCY2C, His Tag (Cat. No. GUC-H52H5) and negative control protein respectively, washed and then followed by PE anti-His antibody and analyzed with FACS (Routinely tested).
  • Gene ID:
    P25092-1
  • Protein synonyms:
    GUCY2C|hSTAR|STA receptor|GC-C|STAR|GUC2C
  • Protein/peptide name:
    GUCY2C
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    47.9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-718

Specifications

About this item

Human GUCY2C / Guanylyl cyclase C Protein, His Tag, ACROBiosystems