Human Recombinant FOLR1 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Catalog # 103871-490
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Size:200 µg
- Tag sequence:His Tag (MALS verified)
- Storage conditions:–20 ℃, aviod light
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:2e5 of Anti-FOLR1 CAR-293 cells were stained with 100 μL of 3 μg/mL of FITC-Labeled Human FOLR1, His Tag (Cat. No. FO1-HF2H8) and negative control protein respectively, FITC signals was used to evaluate the binding activity (QC tested).
- Gene ID:P15328-1
- Protein synonyms:FOLR-1|FOLR|FBP
- Protein/peptide name:FOLR1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:26.5 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-490
Specifications
About this item
FITC-Labeled Human FOLR1 Protein, His Tag (MALS verified), ACROBiosystems