Human Recombinant EphA4 (from HEK293 cells)
103814-962
:
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Human EphA4, His Tag (Cat. No. EP4-H52H8) at 5 μg/mL (100 μL/well) can bind Human Ephrin-A4, Fc Tag (Cat. No. EA4-H5250) with a linear range of 0.6-5 ng/mL (QC tested).
- Gene ID:P54764-1
- Protein synonyms:EK8|TYRO1|HEK8|SEK|EphA4
- Protein/peptide name:EphA4
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:60.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103814-962
Human EphA4 Protein, His Tag, ACROBiosystems