Human Recombinant CD7 (from HEK293 Cells)
Catalog # 103815-164
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Tag sequence:Llama IgG2b Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0.01 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Human CD7, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD7-H5258) at 5 μg/mL (100 μL/well) can bind Human SECTM1, His Tag (Cat. No. SE1-H5227) with a linear range of 10-39 ng/mL (QC tested).
- Gene ID:P09564-1
- Protein synonyms:LEU-9|TP41|CD7|Tp40|GP40
- Protein/peptide name:CD7
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:44.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103815-164
Specifications
About this item
Human CD7 Protein, Llama IgG2b Fc Tag, low endotoxin, ACROBiosystems