Order Entry
Canada
ContactUsLinkComponent
 
HCoV-HKU1(isolate N5) Recombinant S1 protein (from HEK293 cells)
Catalog # 103871-302
CAS Number:  
HCoV-HKU1(isolate N5) Recombinant S1 protein (from HEK293 cells)
Catalog # 103871-302
Supplier Number:  SINV52H6100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Size:
    100 µg
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    Q0ZME7-1
  • Protein synonyms:
    Spike glycoprotein Subunit1|Spike protein S1, COVID-19|S glycoprotein Subunit1|Spike, S1 protein
  • Protein/peptide name:
    S1 protein
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    85.7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103871-302

Specifications

About this item

HCoV-HKU1(isolate N5) S1 protein, His Tag, ACROBiosystems