Cynomolgus Monkey Recombinant Mesothelin (from HEK293 cells)
Catalog # 103871-496
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:A0A2K5TW94-1
- Protein synonyms:MPF|MSLN|Mesothelin
- Protein/peptide name:Mesothelin
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:33.7 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-496
Specifications
About this item
Cynomolgus Mesothelin / MSLN (296-580) Protein, His Tag (MALS verified), ACROBiosystems