Cynomolgus monkey Recombinant CD73 (from HEK293 cells)
Catalog # 103871-048
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Protein synonyms:5'-Nucleotidase|5'-NT|NTE|NT5|NT5E|CD73
- Protein/peptide name:CD73
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:60.0 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103871-048
Specifications
About this item
Cynomolgus CD73 Protein, His Tag (active enzyme), ACROBiosystems