Order Entry
Canada
ContactUsLinkComponent
Cynomolgus monkey Recombinant B7-H2 (from HEK293 cells)
Cynomolgus monkey Recombinant B7-H2 (from HEK293 cells)
Catalog # 103815-110
CAS Number:  
Cynomolgus monkey Recombinant B7-H2 (from HEK293 cells)
Catalog # 103815-110
Supplier Number:  B72C52HB100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Size:
    100 µg
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human ICOS, Fc Tag (Cat. No. ICS-H5258) at 1 μg/mL (100 μL/well) can bind Cynomolgus B7-H2, His Tag (Cat. No. B72-C52Hb) with a linear range of 0.6-20 ng/mL (QC tested).
  • Protein synonyms:
    ICOS-L|ICOS ligand|B7-H2|B7RP-1|ICOSLG|CD275|LICOS|B7RP1|ICOSL|GL50|B7H2
  • Protein/peptide name:
    B7-H2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    28.5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103815-110

Specifications

About this item

Cynomolgus B7-H2 / ICOSLG Protein, His Tag, ACROBiosystems