Cynomolgus monkey Recombinant B7-H3 (from HEK293 cells)
Catalog # 103815-136
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:100 µg
- Tag sequence:His Tag (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus B7-H3, His Tag (Cat. No. B73-C52Ha) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.1-4 ng/mL (QC tested).
- Protein synonyms:B7 homolog 3|B7-H3|CD276
- Protein/peptide name:B7-H3
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:49.0 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103815-136
Specifications
About this item
Cynomolgus B7-H3 / CD276 Protein, His Tag (MALS verified), ACROBiosystems