About this item
This specialty formulated blend of vinyl and nitrile provides a cost effective alternative to the traditional nitrile examination glove, while maintaining the performance expected from a disposable examination glove.
- Lightweight nitrile/vinyl blend provides a sensitive feel
- Latex-free
- Cost competitive solution when compared to nitrile examination gloves
- Easy donning with beaded cuff
- Applicable for many industries
Suitable alternative for individuals with Type I allergies. Great for a wide range of applications including medical, examination, healthcare, nursing, laboratory, IT, general cleaning, and industrial use.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76456-892
- 76456-896
- 76456-894
- 76456-898
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Specifications
Related Information
Certificates
Search by Lot NumberCoQ VWR® Vinyl Nitrile Hybrid Examination Gloves
Conformity declaration
VWR Disposable Glove VWR Glove Chemical Resistance Chart VWR® Disposable Glove Guide VWR® Focus Production Volume 3
Product Literature
VWR Disposable Gloves