Spigot, LDPE
Spigot, LDPE, 3.3×1.15×2.8", 3/4"
Catalog # 470330-666
Supplier: Dynalon
Supplier #: 612935
Jump to:
Product Details & Documents
Standard drum spigot.
Plastic spigots are molded of low-density polyethylene of white with 3/4" IPS thread. Plastic spigots have no gaskets or inside threads to leak. Will fit standard threaded drums.
Specifications
- Description:Spigot
- Material:LDPE
- L×W×H:3.3×1.15×2.8"
- Thread:3/4"