Order Entry
Canada
ContactUsLinkComponent
SP Wilmad-LabGlass Stainless Steel Micro Spatula For NMR and EPR, SP Industries
SP Wilmad-LabGlass Stainless Steel Micro Spatula For NMR and EPR, SP Industries
Catalog # 76459-580
Supplier:  SP Industries
SP Wilmad-LabGlass Stainless Steel Micro Spatula For NMR and EPR, SP Industries
Catalog # 76459-580
Supplier:  SP Industries
Supplier Number:  806

Specifications

  • Description:
    Micro spatula
  • Length:
    250 mm
  • Cat. no.:
    76459-580

Specifications

About this item

Make solid-state and gel-phase NMR sample trasfers easier.

  • Fits into most 4 mm and larger Ext.Ø. tubes
  • Manufactured from 18/8 stainless steel