If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Engineering a drug delivery system
- Cat. no.:470328-172
- No. of Uses:15
Specifications
About this item
Learn about drug delivery systems, which can control the transport and release of medicine in the body.
- Students will gain an understanding of drug delivery and its importance by testing simulated delivery systems in multiple environments that mimic different parts of the body.
- Aligned to NGSS Practices
- Performance expectations: MS-ETS1-1, MS-ETS1-2, MS-LS1-3, HS-ETS1-2
- Disciplinary core ideas: EST1.A, ETS1.B, LS1.A, ETS1.C
- Cross cutting concepts include influence of science enginerring and technology on society and the natural world, systems and system models, science is a human endeavor
- Engineering practices include asking questions and defining problems, engaging in argument from evidence, constructing explanations and designing solutions
Biomedical engineers design devices, equipment, and processes to improve medical treatment methods and the quality of human health and life. They apply engineering principles to biology and medicine to create products such as prosthetic limbs and artificial organs, among others. One major focus of biomedical engineers is the design of drug delivery systems, which can control the transport and release of medicine in the body to more effectively treat diseases and illnesses. In this experiment, students will gain an understanding of drug delivery and its importance by testing simulated delivery systems in multiple environments that mimic different parts of the body. The delivery systems will be analyzed and compared, allowing students to engineer an optimal solution and learn what properties must be considered when designing new drug delivery systems. This lab has enough materials for 15 groups. Teacher's manual and student guide included.