ContactUsLinkComponent
Distance Learning: Mohs Hardness
  470330-518
undefined
Distance Learning: Mohs Hardness
  470330-518
 :  IS1512
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Subjects:
    Earth Science
  • Description:
    Distance Learning: Mohs Hardness
  • No. of Students:
    1
  • Cat. no.:
    470330-518
Determine the identity of minerals.
  • Aligned to the Next Generation Science Standards
  • Contains Instruction Manual
  • Materials for only two groups of students or a single student
The Mohs hardness scale was invented by Friedrich Mohs in 1812, and has been a useful tool for geologists to determine the identity of different minerals. With this kit, students will become familiar with the Mohs hardness scale and how to perform the hardness test, allowing them to develop vital skills that will help them throughout their studies in earth science.
This kit also provides students an opportunity to go out into their local environment to investigate the relative hardness of the minerals around them.