96-Well Polypropylene Storage System, Costar, Cole-Parmer
Catalog # MSPP-VV0172880
Supplier: Cole Parmer
Specifications
- Well Volume:0.32 mL
- Cat. no.:MSPP-VV0172880
- Bottom Style:V-bottom
Specifications
About this item
Standard microplates are built to industry standard footprint, compatible with automated systems.
- Certified RNase- and Dnase-free
Constructed of clarified polypropylene for optimal clarity and solvent resistance which is ideal for compound library and combinatorial chemistry applications.