ContactUsLinkComponent
Chromogenic Analysis of Water Contaminants
Chromogenic Analysis of Water Contaminants
  470107-598
 :  Edvotek
undefined
Chromogenic Analysis of Water Contaminants
  470107-598
 :  Edvotek
 :  951
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Chromogenic Analysis of Water Contaminants
  • No. of Lab Groups:
    10
  • Cat. no.:
    470107-598
  • Supplier No.:
    951
Test water from your own environment for fecal coliforms.
  • Complete assay in 30 minutes, and grow overnight
  • Non-pathogenic bacteria provided
  • Easy to interpret results
  • Includes instructor's guide and discussion questions
In this experiment, students will test control and environmental water samples for the presence of total coliform bacteria and E. coli using color changing enzyme assays. Testing drinking water for every possible type of pathogenic bacteria is slow and costly. Thus, drinking water is tested for coliforms - including the familiar E. coli. Presence of coliforms is an indicator of fecal contamination.
Bactobead refill contains positive control organisms: Escherichia coli, and Citrobacter freundii.
 : Additional equipment needed but not included: Long wave UV Lamp, 10% bleach solution or other disinfectant, water-collection bottles (e.g. screw-cap drink bottles). Water bath or incubator and micropipettes are helpful but not required.
 : Items will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment.
 : Some components require Refrigerated storage upon receipt.