MZone™ FlatMop™ Slip-On Mop Covers, Micronova
These slip-on, tab removal flat head mop covers are ideal for cleaning and sanitizing all surfaces and equipment in the cleanroom. A choice of fabric options provide excellent absorbency, effective cleaning, and disinfectant dispersal for specialized cleaning.
- Easy to change covers fit securely over mop frame
- Urethane foam interiors fully encased in a choice of fabric:
- NovaPoly - 100% smooth knit polyester provides excellent disinfectant dispersal
- PolySorb - 100% textured polyester for removing residues and dispersing disinfectant
- MegaTex - 100% non-woven polyamide is absorbent with a mildly abrasive surface
- Microfiber - 80% polyester or 20% nylon for maximum absorbency and residue removal
- PolyMesh -- 100% polyester with an abrasive mesh layer for exellent residue removal
- The 9" covers are excellent for pass-throughs and smaller spaces
- The 14 and 18" covers are ideal for floors, walls, and ceilings
The Micronova MZone™ FlatMop™ Mop covers fit onto the 360° swivel SlimLine™ adapter, which can be locked at 180° for more controlled cleaning. Available gamma irradiated for aseptic environments. Suitable for use with strong disinfectants and repeated autoclaving.
Connection: Slip-on, tab removal
Interior: Urethane foam
- Catalog No:
- 76383-048
- 10153-676
- 76472-548
- 76383-042
- 76383-046
- 76383-044
- 89498-080
- 89498-078
- 10153-674
- 89498-076
- 89498-074
- 76214-288
- 76383-038
- 75848-692
- 75848-694
- 76214-286
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
SlimLine™ Plastic Mop Adapters
Frequently Bought Together