ContactUsLinkComponent
Eisco Polypropylene Measuring Jugs, Screen Printed
Eisco Polypropylene Measuring Jugs, Screen Printed
Catalog # 470328-324
Supplier:  Wards
Supplier #: CH0357A
undefined
Eisco Polypropylene Measuring Jugs, Screen Printed
Catalog # 470328-324
Supplier:  Wards
Supplier Number:  CH0357A
Supplier #: CH0357A
Log in for availability
Select an Option
10 ml
Select an Option
100 ml
Select an Option
470328-324
About this item
Easy to read and easy to pour shatterproof measuring jug with screen printed graduations.
  • Made with high quality polypropylene
  • Features easy to read scree printed graduations
  • Excellent clarity
  • Autoclavable
  • Good chemical resistance
  • Single piece construction with handle for easy carrying and lifting
  • No drip spout for mess free pouring
Eisco Labs manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Frequently Bought Together