HAZWIK® Quick Response Kit – Bodily Fluids, Brady Worldwide
Catalog # 76458-742
Supplier: Brady Worldwide
Specifications
- Description:Quick response spill kit, bodily fluids
- Dimensions:19×9×13.125"
- Size:0.9 lbs. bag
- Cat. no.:76458-742
Specifications
About this item
Get fast and effective spill cleanup with a HAZWIK® Quick Response Kit. It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.
- Quickly cleans up hazardous bodily fluid spills: blood, vomit and urine
- Visual instructions on the package
Compact, single-use kit ideal for use in schools, hospitals and on janitorial carts. Contains PPE, tools, absorbent pads, organic granular, wipes and disposable materials.
Delivery: Includes: 6× 7.5×10" pads; 1× pair nitrile gloves; 1× bag organic coconut coir granular; 1× scoop and scrapper; 1× disposal bag; 1× cable tie; 1× surface sanitizing wipe; 1× hand sanitizing wipe.
Packaging: Sold in packs of 5 kits. Pallet quantity is 250.