Order Entry
Canada
ContactUsLinkComponent
HAZWIK® Quick Response Kit, Chemical, Brady Worldwide
HAZWIK® Quick Response Kit, Chemical, Brady Worldwide
Catalog # 76458-740
Supplier:  Brady Worldwide
undefined
HAZWIK® Quick Response Kit, Chemical, Brady Worldwide
Catalog # 76458-740
Supplier:  Brady Worldwide
Supplier Number:  SKHAZ-QRK-C5

Specifications

  • Description:
    Quick response spill kit, chemical
  • Dimensions:
    19×9×13.125"
  • Size:
    1.2 lbs. bag
  • Cat. no.:
    76458-740

Specifications

About this item

Get fast and effective spill cleanup with a HAZWIK® Quick response kit. It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.

  • Quickly cleans up hazardous chemical spills
  • Visual instructions on the package

Compact, single-use kit that is ideal for use on small spills in the lab. Contains PPE, tools, absorbent pads, acid and base neutralizers, wipes and disposable materials.

Delivery: Includes: 6× 7.5×10" pads; 1× 4.5 oz. acid neutralizer; 1× 4.5 oz base neutralizer; 1× pair nitrile gloves; 1× scoop and scrapper; 1× disposal bag; 1× cable tie; 1× surface sanitizing wipe.
Packaging: Sold in packs of 5 kits. Pallet quantity is 250.