Order Entry
Canada
ContactUsLinkComponent
Staphylococcus aureus V8 Protease
Staphylococcus aureus V8 Protease
Catalog # CAMB-114-0001
CAS Number:  
Staphylococcus aureus V8 Protease
Catalog # CAMB-114-0001
Supplier Number:  MB-114-0001
CAS Number:  

Specifications

  • Species:
    Staphylococcus aureus V8
  • Size:
    1 mg
  • Enzyme name:
    Protease
  • Cat. no.:
    CAMB-114-0001
  • Supplier No.:
    MB-114-0001

Specifications

About this item

All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.

Application: Cleaving peptide bonds on the COOH-terminal side of either aspartic or glutamic acids.