Order Entry
Canada
ContactUsLinkComponent
Paenibacillus polymyxa Neutral Protease Dispase
Paenibacillus polymyxa Neutral Protease Dispase
Catalog # CAMB-127-0050
CAS Number:  
Paenibacillus polymyxa Neutral Protease Dispase
Catalog # CAMB-127-0050
Supplier Number:  MB-127-0050
CAS Number:  

Specifications

  • Species:
    Paenibacillus polymyxa
  • Size:
    50 mg
  • Enzyme name:
    Neutral Protease Dispase
  • Cat. no.:
    CAMB-127-0050
  • Supplier No.:
    MB-127-0050

Specifications

About this item

All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.

Application: Separate skin epidermis from dermis leaving intact epithelial sheets and stem cell, hepatocyte and other cell isolation applications.