Bovine Ribonuclease A, DNase- and Protease-Free (from Pancreas)
Supplier: Rockland Immunochemical
Certificates
About this item
All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.
Applications include, purifying DNA plasmids, removing RNA from DNA, or any type of nucleic acid research.
Specifications
- Source:Pancreas
- Species:Bovine
- Enzyme name:Ribonuclease A, DNase- and Protease-Free
- Protease-free:Y
Specifications
Frequently Bought Together