High Performance FR Knit Men's Button Up Shirts, Khaki, Lakeland Industries
: Lakeland
Featuring Lakeland’s lightweight knit fabric offering permanent moisture-wicking technology. This is the optimal blend for superior performance and protection and comfort.
- 6.9 oz. lightweight inherently FR fabric
- Permanent moisture wicking properties
- Tag-free design
- Button down collar
- Athletic cut
- Two chest pockets
Mid layers are the versatile garments that can help take the chill off for a few hours on a fall morning or provide an extra layer of insulation for those long, frigid winter days. Paired with a Base layer, Mid layers continue to pull moisture away from the body and move it to the outermost garment so it can dry. Lightweight, effective fabrics let Mid layers increase comfort without reducing mobility.
: NFPA 70E. NFPA 2112 - UL certified. APTV: 16.4 cal/cm². CAT: 2.
- Catalog No:
- 76438-746
- 76438-756
- 76438-758
- 76438-752
- 76438-754
- 76438-764
- 76438-766
- 76438-744
- 76438-760
- 76438-762
- 76438-748
- 76438-770
- 76438-750
- 76438-768
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- FR fabric
- Weight:
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
- 6.9 oz.
Frequently Bought Together