To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:10 per pack
- Description:Prepared media (BEA with Azide/CNA agar)
- Size:15×100 mm
- Cat. no.:76441-340
- Type:Biplate
Specifications
About this item
Bile esculin agar with azide / columbia agar with 5% sheep blood, colistin and nalidixic acid for the presumptive identifcation of group D streptococci including enterococci and gram positive bacteria.
This Columbia CNA agar is recommended for use as a selective growth medium for the isolation and differentiation of Gram-positive cocci from clinical and non-clinical specimens which contain mixed flora.