ContactUsLinkComponent
Dominator™ DM3 Series Safety Glasses, MCR Safety
  76342-916
undefined
Dominator™ DM3 Series Safety Glasses, MCR Safety
  76342-916
 :  DM1310PF
  • Frame Color:
    Gunmetal/red
  • Description:
    Safety glasses VLT 90%
  • Cat. no.:
    76342-916
  • Lens Coating:
    Anti-fog (MAX6™)
  • Lens Tint:
    Clear
  • Frame Material:
    PC
The Dominator™ DM3 Series is one of the most versatile and adjustable safety glasses available, featuring wire core temples which allow the user to bend or adjust each temple to create a customized fit and feel.
  • Italian fashion, sport focused design
  • Co-injected wire core independently adjustable nose pads
  • Co-injected soft wire core adjustable TPR temples
  • MAX6™ anti-fog coating is proven to provide six times greater fog-resistance and dissipation
These high-quality safety glasses have a lifetime frame warranty and are engineered to provide the highest level of innovation.
 : ANSI Z87+. MIL - PRF - 31013.