ContactUsLinkComponent
UD 1 Series Safety Glasses, MCR Safety
  76343-354
undefined
UD 1 Series Safety Glasses, MCR Safety
  76343-354
 :  UD110PF
  • Frame Color:
    Gunmetal
  • Description:
    Safety glasses VLT 90%
  • Cat. no.:
    76343-354
  • Lens Coating:
    Anti-fog (MAX6™)
  • Lens Tint:
    Clear
  • Frame Material:
    PC
UD1 Safety glasses are distinguished with application based, stylish design elements and superior coatings for work and play.
  • Entire series meets military ballistic high velocity standard
  • MAX6™ anti-fog coating is proven to provide six times greater fog-resistance
  • Sleek co-injected TPR over polycarbonate temples
  • Fixed closed cell foam lined options available
  • Modern stylish design
The sleek and stylish modern design features non-slip temple grips, soft TPR nose pads, and crisp bayonet temples. These high-quality safety glasses have a lifetime frame warranty and are engineered to provide the highest level of innovation.
 : ANSI Z87+. MIL - PRF - 31013.