Order Entry
Canada
ContactUsLinkComponent
 
Monarch® RNA Cleanup Kits (50 μg), New England Biolabs
 
undefined
Monarch® RNA Cleanup Kits (50 μg), New England Biolabs

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified RNA
  • Sample type:
    RNA preps
    PCR and other enzymatic reactions
  • Target molecule:
    RNA
  • Description:
    Monarch® RNA cleanup kit

 

Columns

 

Frequently Bought Together