ContactUsLinkComponent
Monarch® RNA Cleanup Columns, New England Biolabs
Monarch® RNA Cleanup Columns, New England Biolabs
Catalog # CA76307-362
CAS Number:  
Monarch® RNA Cleanup Columns, New England Biolabs
Catalog # CA76307-362
Supplier Number:  T2037L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Monarch® RNA Cleanup Columns, 10 μg/100 columns
  • Cat. no.:
    CA76307-362
Specifications
About this item
The Monarch® RNA cleanup columns (10 µg) are a component of the Monarch RNA cleanup kit (10 µg) and can be used to purify up to 10 µg of RNA from enzymatic reactions. They are supplied with Monarch® collection tubes II. RNA cleanup binding buffer and Monarch® RNA wash buffer, are also available separately.
  • Optimal for cleaning up RNA from a variety of enzymatic reactions
  • Ideal for the concentration of RNA into small volumes (elute in ≥6 µl)
  • 70 to 100% RNA recovery, even with inputs <20 ng
  • Purified RNA is ready for use in a wide variety of downstream applications
  • Unique column design prevents buffer carryover and elution of silica particulates
The columns are designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants. RNA can be eluted in as little as 6 μl and is ready for use in a variety of downstream applications including RT-PCR, RNA library prep for NGS, and labeling.
Monarch® RNA cleanup columns are also available for 50 and 500 µg binding capacities.