Linearity FLQ Anemia Siemens Centaur, AUDIT Microcontrols
Catalog # 76446-936
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity FLQ anemia Siemens Centaur
- Size:5×2 ml
- Cat. no.:76446-936
- Stability:7 days when stored at 2...8 °C
- Analyte:Cortisol, Ferritin, Folate, Vitamin B12
- Type:Frozen liquid
Specifications
About this item
Linearity FLQ anemia for Siemens centaur is an IVD device consisting of five levels.
- Format: frozen liquid
- 5 levels
It is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: cortisol, ferritin, folate, vitamin B12.