Order Entry
Canada
Orders LinkContactUsLinkComponent
Performance Check Solution for US EPA Method 508, SPEX CertiPrep

Performance Check Solution for US EPA Method 508, SPEX CertiPrep

Performance Check Solution for US EPA Method 508, 1 ml, Methyl tertiary-butyl ether:Ethyl acetate, Ampoule, amber
Catalog # SX5081-PC
Supplier:   SPEX CERTIPREP LLC
Supplier #:  5081-PC
Performance Check Solution for US EPA Method 508, SPEX CertiPrep
Catalog # SX5081-PC
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Performance Check Solution for US EPA Method 508, SPEX CertiPrep

Performance Check Solution for US EPA Method 508, 1 ml, Methyl tertiary-butyl ether:Ethyl acetate, Ampoule, amber

CAS Number:  
Price
$96.64
Log in for availability
Each (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Pesticides, PCBs and Herbicides

The EPA is tasked with the monitoring of environmental systems as they pertain to contamination and human health. The methods issued by the EPA are created to respond to specific toxins or persistent organic pollutants (POPs) found in environmental samples such as soil, source water, drinking water, and waste. In particular, pollutants such as polychlorinated biphenyls (PCBs), which were in widespread industrial use up until their restriction, are of concern due to their stable and persistent nature in the environment. Another group of chemicals of high concern is the hundreds of commercial pesticides and herbicides in use in the world today. Pesticides, from algaecides to virucides, are used in large quantities in industrial and private agriculture. The concern over human pesticide exposure over the past few decades has led to extensive monitoring of these pesticides. It has been reported that over 98% of insecticides and 95% of herbicides affect areas other than their intended target product. It is essential that monitoring agencies have accurate standard mixes to measure the pesticide levels in the environment.

Many new pesticides are now being tested using highly sensitive LC/MS techniques, in addition to traditional GC techniques, to determine minute amounts of residue in environmental samples and food products.

At SPEX CertiPrep, we facilitate ease of monitoring and testing of pesticides by creating pesticide test mixes to suit your monitoring needs. SPEX CertiPrep is the leader in offering pesticide standards designed to work within EPA, AOAC and FDA analytical testing methods using all of the leading analytical techniques: LC, LC/MS, GC, and GC/MS. Many pesticide standard mixes are readily available in our catalog along with a large list of single pesticide standards. In addition, custom pesticide mixes can be made to your specifications to create a mix that meets your needs.

US EPA Method 508 is an analytical method for the monitoring of organochlorine pesticides and PCBs in drinking water and ground water by GC/ECD

Documents

Specifications

Specifications

  • Matrix:
    Methyl tertiary-butyl ether:Ethyl acetate
  • Pack type:
    Ampoule, amber
  • Description:
    Performance Check Solution for US EPA Method 508
  • Size:
    1 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.