n-Hexane, SPEX CertiPrep
Product Details & Documents
n-Hexane is an analytical standard that can be used for many applications including petroleum and LUST standards, residual solvent standards for medicinal and recreational cannabis testing and also for pollutants in wastewater and water testing.
The Hazardous and Solid Waste Amendments to the Resource Conservation and Recovery Act (RCRA), passed in 1984, required the US Environmental Protection Agency (EPA) to develop regulations for the underground storage of motor fuels. The EPA requires owners and operators of USTs to maintain and clean up sites damaged by petroleum contamination.
Specifications
Specifications
- Concentration:1000 µg/ml
- Matrix:Methanol (P&T)
- Pack type:Ampoule, amber
- Description:n-Hexane
- Size:1 ml
- Cat. No.:SXS-2190
- Description:n-Hexane
- CAS No.:110-54-3
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



