Order Entry
Canada
ContactUsLinkComponent
Acid Extractables Spiking Mix for US EPA Methods 625 and 8270, SPEX CertiPrep
Acid Extractables Spiking Mix for US EPA Methods 625 and 8270, SPEX CertiPrep
Catalog # SXECS-N-004
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Acid Extractables Spiking Mix for US EPA Methods 625 and 8270, SPEX CertiPrep
Catalog # SXECS-N-004
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  ECS-N-004
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    2000 µg/ml
  • Matrix:
    Methylene chloride
  • Pack type:
    Glass bottle, amber
  • Description:
    Acid Extractables Spiking Mix for US EPA Methods 625 and 8270
  • Size:
    5.5 ml
  • Environmentally Preferable:
  • Cat. no.:
    SXECS-N-004

Specifications

About this item

ECS Standards for GC/MS

All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.