Order Entry
Canada
Orders LinkContactUsLinkComponent
Method C1001-09.4 Phthalates, SPEX CertiPrep

Method C1001-09.4 Phthalates, SPEX CertiPrep

Method C1001-09.4 Phthalates, 1 ml, 1000 µg/ml, Isooctane, Glass bottle, amber
Catalog # SXC1001-09.4
Supplier:   SPEX CERTIPREP LLC
Supplier #:  C1001-09.4
Method C1001-09.4 Phthalates, SPEX CertiPrep
Catalog # SXC1001-09.4
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Method C1001-09.4 Phthalates, SPEX CertiPrep

Method C1001-09.4 Phthalates, 1 ml, 1000 µg/ml, Isooctane, Glass bottle, amber

CAS Number:  
Price
$130.23
Log in for availability
Each (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Consumer safety standards, phthalates in polyvinyl chloride (PVC) QC standards.

Polyvinyl chloride, or PVC, is a very common plastic used in a wide range of common consumer products from children’s toys and care items to building and construction materials. In the US, ASTM (American Society for Testing and Materials) and CPSC (Consumer Product Safety Commission) have designed methods for testing children’s toys and childcare articles for compliance with the restriction of use for eight designated phthalates.

Specifications

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Isooctane
  • Pack type:
    Glass bottle, amber
  • Description:
    Method C1001-09.4 Phthalates
  • Size:
    1 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.