Order Entry
Canada
ContactUsLinkComponent
Trichloroethene Single-Component Organic Standard, SPEX CertiPrep
Trichloroethene Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-3615
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Trichloroethene Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-3615
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  S-3615
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methanol (P&T)
  • Pack type:
    Ampoule, amber
  • CAS No.:
    79-01-6
  • Description:
    Trichloroethene
  • Size:
    1 ml
  • Environmentally Preferable:
  • Description:
    Trichloroethene
  • Cat. no.:
    SXS-3615

Specifications

About this item

Consumer safety standards. Pharmaceutical residual solvent standards.

In the pharmaceutical industry, the guidelines set by the International Conference on Harmonization (ICH), and by United States Pharmacopeia (USP) and European Pharmacopeia (EP), mandate that manufacturing solvents have to be regulated due to their toxic and/or environmentally hazardous nature.