Order Entry
Canada
ContactUsLinkComponent
 
Canadian Cannabis Residual Solvent Mix 1, SPEX CertiPrep
Catalog # SXCAN-CAN-RS-1
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Canadian Cannabis Residual Solvent Mix 1, SPEX CertiPrep
Catalog # SXCAN-CAN-RS-1
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CAN-CAN-RS-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methanol (P&T)
  • Pack type:
    Glass bottle, amber
  • Description:
    Canadian Cannabis Residual Solvent Mix 1
  • Size:
    1 ml
  • Cat. no.:
    SXCAN-CAN-RS-1

Specifications

About this item

Cannabis standard, analytical standard for medicinal and recreational cannabis testing.
Designed for Methods: State specific pesticide regulations:

  • OAR 333-008-11
  • HB 3460
  • AOAC 2007-01
  • EN 15662

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

SPEX CertiPrep offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.