Order Entry
Canada
ContactUsLinkComponent
Sodium thiosulfate 0.0394 N, Carbon Black reagent for ASTM D1510
Sodium thiosulfate 0.0394 N, Carbon Black reagent for ASTM D1510
Catalog # SX182002
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Sodium thiosulfate 0.0394 N, Carbon Black reagent for ASTM D1510
Catalog # SX182002
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  182002
CAS Number:  

Specifications

  • Pack type:
    Cubitainer®
  • Size:
    1 gal.
  • Cat. no.:
    SX182002

Specifications

About this item

 :
 :158.11 g/mol
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499