Magnesium (Mg²⁺) Single-Element Ion Cation Standard, SPEX CertiPrep
SXCS-MG2-2Y
:
- Concentration:1000 µg/ml
- Matrix:0.2% HNO₃
- Pack type:LDPE bottle
- Size:125 ml
- Cat. no.:SXCS-MG2-2Y
- Element:Magnesium
Single-element standards for ion chromatography and ion selective electodes.
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.