Lithium (Li+) Single-Element Ion Cation Standard, SPEX CertiPrep
Catalog # SXCS-LI2-2Y
Supplier: SPEX CERTIPREP LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml
- Matrix:0.2% HNO₃
- Pack type:LDPE bottle
- Size:125 ml
- Cat. no.:SXCS-LI2-2Y
- Element:Lithium
Specifications
About this item
Single-element standards for ion chromatography and ion selective electodes.
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.